Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Alpi
Entrez Gene ID
76768
Gene Name
alkaline phosphatase, intestinal
Gene Aliases
2010001C14Rik, Akp6, gIAP
Chromosome Location
Chr.1: 87098002 - 87101606 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001081082.2 NP_001074551.1 10 - 11 1374 60
GenBank mRNA AK008000.1 - 4 - 5 464 60
BC119800.1 - 10 - 11 1302 60

View Full Product Details