Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Mlph
Entrez Gene ID
171531
Gene Name
melanophilin
Gene Aliases
2210418F23Rik, 5031433I09Rik, AW228792, D1Wsu84e, Slac-2a, l(1)-3Rk, l1Rk3, ln
Chromosome Location
Chr.1: 90915022 - 90951142 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_053015.3 NP_443748.2 11 - 12 1584 85
GenBank mRNA AB057759.1 - 10 - 11 1406 85
AF384098.1 - 11 - 12 1581 85
AK019883.1 - 11 - 12 1555 85
AK148274.1 - 11 - 12 1556 85
BC006900.1 - 11 - 12 1577 85
BC150804.1 - 11 - 12 1478 85

View Full Product Details