The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001159965.1 | NP_001153437.1 | 12 - 13 | 1276 | 75 |
NM_001159966.1 | NP_001153438.1 | 12 - 13 | 1174 | 75 | |
NM_001159967.1 | NP_001153439.1 | 12 - 13 | 1276 | 75 | |
NM_001159968.1 | NP_001153440.1 | 11 - 12 | 1171 | 75 | |
NM_023884.4 | NP_076373.3 | 12 - 13 | 1216 | 75 | |
NR_027640.1 | 13 - 14 | 1381 | 75 | ||
GenBank mRNA | AF312924.1 | - | 11 - 12 | 1124 | 75 |
AK018622.1 | - | 12 - 13 | 1218 | 75 | |
AK019543.1 | - | 11 - 12 | 1174 | 75 | |
AK029049.1 | - | 12 - 13 | 1217 | 75 | |
AK029993.1 | - | 13 - 14 | 1354 | 75 | |
AK033549.1 | - | 12 - 13 | 1172 | 75 | |
AK036803.1 | - | 11 - 12 | 1148 | 75 | |
AK161080.1 | - | 12 - 13 | 1243 | 75 | |
AK165002.1 | - | 12 - 13 | 1168 | 75 | |
AK169368.1 | - | 12 - 13 | 1194 | 75 | |
BC043132.1 | - | 12 - 13 | 1181 | 75 | |
BC052663.1 | - | 12 - 13 | 1193 | 75 |