Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
4930555M17Rik
Entrez Gene ID
78039
Gene Name
RIKEN cDNA 4930555M17 gene
Gene Aliases
-
Chromosome Location
Chr.3: 5512995 - 5515376 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
GenBank mRNA AK019767.1 - 1 - 2 171 67


Non Gene Transcripts

Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Transcript Location Amplicon Length
GenBankmRNA BY715739.1 - 1 - 2 171 Chr. 3 - 5512993 - 5515333 [-] on Build GRCm38 67


Adjacent Gene Details

Gene Name Gene Symbol Alias Distance Relative Direction Location Chromosome
Upstream zinc finger homeodomain 4 Zfhx4 A930021B15, C130041O22Rik, Zfh-4, Zfh4 -97.1kb Anti-sense Chr. - 5217420 - 5415861 on Build GRCm38
Downstream peroxisomal biogenesis factor 2 Pex2 D3Ertd138e, PAF-1, PMP35, Pxmp3 44.9kb Sense Chr. - 5560188 - 5576877 on Build GRCm38

View Full Product Details