Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Rsad2
Entrez Gene ID
58185
Gene Name
radical S-adenosyl methionine domain containing 2
Gene Aliases
2510004L01Rik, Vig1, cig5
Chromosome Location
Chr.12: 26442743 - 26456478 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
GenBank mRNA AF236064.2 - 2 - 3 520 69
AF442152.1 - 2 - 3 505 69

View Full Product Details