Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Inpp5d
Entrez Gene ID
16331
Gene Name
inositol polyphosphate-5-phosphatase D
Gene Aliases
SHIP, SHIP-1, SHIP1, SIP-145, p150Ship, s-SHIP
Chromosome Location
Chr.1: 87620312 - 87720510 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001110192.2 NP_001103662.1 13 - 14 1740 93
NM_001110193.2 NP_001103663.1 13 - 14 1743 93
NM_010566.3 NP_034696.2 13 - 14 1743 93
NR_122078.1 13 - 14 1740 93
GenBank mRNA AF125996.1 - 13 - 14 1656 93
AF184912.1 - 8 - 9 936 93
AF184913.1 - 8 - 9 936 93
AF228679.1 - 13 - 14 1654 93
AK143560.1 - 13 - 14 1680 93
BC108328.1 - 13 - 14 1616 93
U39203.1 - 13 - 14 1692 93
U51742.1 - 13 - 14 1654 93

View Full Product Details