The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_172844.2 | NP_766432.1 | 8 - 9 | 1392 | 78 |
GenBank mRNA | AK029219.1 | - | 8 - 9 | 1393 | 78 |
AK161224.1 | - | 8 - 9 | 1392 | 78 | |
BC111885.1 | - | 6 - 7 | 1121 | 78 | |
BC112405.1 | - | 8 - 9 | 1382 | 78 |