The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_172844.2 | NP_766432.1 | 9 - 10 | 1464 | 65 |
GenBank mRNA | AK029219.1 | - | 9 - 10 | 1465 | 65 |
AK161224.1 | - | 9 - 10 | 1464 | 65 | |
BC111885.1 | - | 7 - 8 | 1193 | 65 | |
BC112405.1 | - | 9 - 10 | 1454 | 65 |