The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001163154.1 | NP_001156626.1 | 8 - 9 | 1021 | 75 |
NM_007960.4 | NP_031986.1 | 11 - 12 | 1361 | 75 | |
GenBank mRNA | AK032224.1 | - | 8 - 9 | 1023 | 75 |
AK035914.1 | - | 9 - 10 | 1752 | 75 | |
AK144713.1 | - | 11 - 12 | 1310 | 75 | |
BC005645.1 | - | 11 - 12 | 1289 | 75 | |
BE859344.1 | - | 1 - 2 | 136 | 75 | |
L10426.1 | - | 11 - 12 | 1310 | 75 |