Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Pfkfb2
Entrez Gene ID
18640
Gene Name
6-phosphofructo-2-kinase/fructose-2,6-biphosphatase 2
Gene Aliases
4930568D07Rik
Chromosome Location
Chr.1: 130689043 - 130729253 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001162415.1 NP_001155887.1 11 - 12 1225 77
NM_001162416.1 NP_001155888.1 9 - 10 1018 77
NM_008825.4 NP_032851.2 10 - 11 1119 77
NR_027859.1 10 - 11 1119 77
GenBank mRNA AK029659.1 - 11 - 12 1226 77
AK146744.1 - 10 - 11 1113 77
BC018418.1 - 10 - 11 1076 77
BC051014.1 - 9 - 10 1032 77
EU616666.1 - 8 - 9 892 77
EU616667.1 - 9 - 10 1017 77
EU616668.1 - 9 - 10 1017 77
EU616669.1 - 9 - 10 1017 77
EU616670.1 - 8 - 9 883 77
X98847.1 - 9 - 10 1037 77

View Full Product Details