The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001162415.1 | NP_001155887.1 | 4 - 5 | 446 | 71 |
NM_001162416.1 | NP_001155888.1 | 2 - 3 | 239 | 71 | |
NM_008825.4 | NP_032851.2 | 3 - 4 | 340 | 71 | |
NR_027859.1 | 3 - 4 | 340 | 71 | ||
GenBank mRNA | AK029659.1 | - | 4 - 5 | 447 | 71 |
AK146744.1 | - | 3 - 4 | 334 | 71 | |
BC018418.1 | - | 3 - 4 | 297 | 71 | |
BC051014.1 | - | 2 - 3 | 253 | 71 | |
EU616666.1 | - | 2 - 3 | 238 | 71 | |
EU616667.1 | - | 2 - 3 | 238 | 71 | |
EU616668.1 | - | 2 - 3 | 238 | 71 | |
EU616669.1 | - | 2 - 3 | 238 | 71 | |
EU616670.1 | - | 2 - 3 | 248 | 71 | |
X98847.1 | - | 2 - 3 | 258 | 71 |