Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Cog6
Entrez Gene ID
67542
Gene Name
component of oligomeric golgi complex 6
Gene Aliases
4933405E16Rik, AU018618, mKIAA1134
Chromosome Location
Chr.3: 52982123 - 53017223 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_026225.3 NP_080501.2 6 - 7 647 100
GenBank mRNA AB093286.1 - 6 - 7 619 100
AK031149.1 - 6 - 7 641 100
AK045223.1 - 6 - 7 641 100
AK048986.1 - 6 - 7 634 100
AK153320.1 - 6 - 7 636 100
BC025427.1 - 6 - 7 653 100

View Full Product Details