Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Fnip1
Entrez Gene ID
216742
Gene Name
folliculin interacting protein 1
Gene Aliases
A730024A03Rik, AI838773, AW557298
Chromosome Location
Chr.11: 54438146 - 54518241 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_173753.4 NP_776114.2 16 - 17 3422 119
GenBank mRNA AK042779.1 - 3 - 4 887 119
AK047814.1 - 3 - 4 663 119
AK122574.1 - 16 - 17 3370 119
BC024776.1 - 1 - 2 269 119
BC079892.1 - 16 - 17 3412 119

View Full Product Details