Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Nucks1
Entrez Gene ID
98415
Gene Name
nuclear casein kinase and cyclin-dependent kinase substrate 1
Gene Aliases
2700010L10Rik, 8430423A01Rik, AI647518, C78391, Nucks
Chromosome Location
Chr.1: 131910458 - 131936321 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_175294.3 NP_780503.2 5 - 6 655 90
GenBank mRNA AB049825.1 - 5 - 6 634 90
BC039951.1 - 5 - 6 567 90

View Full Product Details