The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001159393.1 | NP_001152865.1 | 4 - 5 | 633 | 73 |
NM_001159396.1 | NP_001152868.1 | 4 - 5 | 588 | 73 | |
NM_008390.2 | NP_032416.1 | 4 - 5 | 633 | 73 | |
GenBank mRNA | AK152005.1 | - | 4 - 5 | 597 | 73 |
AK152104.1 | - | 4 - 5 | 596 | 73 | |
AK152193.1 | - | 4 - 5 | 597 | 73 | |
AK153514.1 | - | 4 - 5 | 597 | 73 | |
AK155983.1 | - | 4 - 5 | 597 | 73 | |
AK157347.1 | - | 4 - 5 | 597 | 73 | |
AY225160.1 | - | 3 - 4 | 396 | 73 | |
BC003821.1 | - | 4 - 5 | 566 | 73 | |
CT010234.1 | - | 3 - 4 | 364 | 73 | |
M21065.1 | - | 4 - 5 | 569 | 73 |