Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Nhp2
Entrez Gene ID
52530
Gene Name
NHP2 ribonucleoprotein
Gene Aliases
2410130M07Rik, D11Ertd175e, Nola2
Chromosome Location
Chr.11: 51619773 - 51623714 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_026631.3 NP_080907.1 2 - 3 278 110
GenBank mRNA AK007340.1 - 2 - 3 266 110
AK008766.1 - 2 - 3 305 110
BC024944.1 - 2 - 3 269 110

View Full Product Details