Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Cdkl3
Entrez Gene ID
213084
Gene Name
cyclin-dependent kinase-like 3
Gene Aliases
AI852479, B230379H01Rik
Chromosome Location
Chr.11: 52000858 - 52089778 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001166653.1 NP_001160125.1 4 - 5 756 86
NM_001166654.1 NP_001160126.1 4 - 5 846 86
NM_001166655.1 NP_001160127.1 4 - 5 849 86
NM_001166656.1 NP_001160128.1 4 - 5 756 86
NM_001166657.1 NP_001160129.1 4 - 5 846 86
NM_153785.4 NP_722480.3 4 - 5 846 86
GenBank mRNA AK045036.1 - 3 - 4 1205 86
AK045356.1 - 4 - 5 746 86
AK046394.1 - 4 - 5 847 86
AK050990.1 - 4 - 5 846 86
AK078804.1 - 4 - 5 738 86
AK166428.1 - 4 - 5 688 86
BC028871.1 - 4 - 5 718 86

View Full Product Details