The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_029004.1 | NP_083280.1 | 4 - 5 | 574 | 64 |
GenBank mRNA | AK018593.1 | - | 4 - 5 | 563 | 64 |
AK039330.1 | - | - | 938 | 64 | |
AK047088.1 | - | 3 - 4 | 453 | 64 |