The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_148673.3 | NP_683514.2 | 16 - 17 | 1765 | 85 |
NM_198936.1 | NP_945174.1 | 15 - 16 | 1686 | 85 | |
GenBank mRNA | AK029117.1 | - | 15 - 16 | 1656 | 85 |
AK046262.1 | - | 15 - 16 | 1657 | 85 | |
AK049178.1 | - | 15 - 16 | 1655 | 85 | |
BC013810.1 | - | 13 - 14 | 1285 | 85 | |
BC023057.1 | - | 5 - 6 | 540 | 85 |