Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Atl1
Entrez Gene ID
73991
Gene Name
atlastin GTPase 1
Gene Aliases
4930435M24Rik, Adfsp, Fsp1, Spg3, Spg3a
Chromosome Location
Chr.12: 69893105 - 69964085 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
GenBank mRNA AK015327.1 - 6 - 7 950 67

View Full Product Details