Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Larp1b
Entrez Gene ID
214048
Gene Name
La ribonucleoprotein domain family, member 1B
Gene Aliases
1700108L22Rik, 4933421B21Rik, AA690246, Larp2
Chromosome Location
Chr.3: 40950631 - 40977797 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
GenBank mRNA AK016857.1 - - 346 66
AK136175.1 - - 368 66

View Full Product Details