Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Exosc9
Entrez Gene ID
50911
Gene Name
exosome component 9
Gene Aliases
PM/Scl-75, Pmscl1, RRP45, p5, p6
Chromosome Location
Chr.3: 36552606 - 36565727 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
GenBank mRNA BC158075.1 - 4 - 5 414 155

View Full Product Details