Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Bbs7
Entrez Gene ID
71492
Gene Name
Bardet-Biedl syndrome 7 (human)
Gene Aliases
8430406N16Rik
Chromosome Location
Chr.3: 36573140 - 36613489 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_027810.3 NP_082086.2 4 - 5 450 75
GenBank mRNA AF521645.1 - 4 - 5 449 75
AK050291.1 - 4 - 5 448 75
AK132661.1 - 4 - 5 378 75
BC031505.1 - 4 - 5 400 75

View Full Product Details