Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Cp
Entrez Gene ID
12870
Gene Name
ceruloplasmin
Gene Aliases
D3Ertd555e
Chromosome Location
Chr.3: 19956987 - 20009750 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001276248.1 NP_001263177.1 13 - 14 2642 112
NM_001276250.1 NP_001263179.1 13 - 14 2642 112
NM_007752.3 NP_031778.2 13 - 14 2645 112
GenBank mRNA AK014835.1 - 13 - 14 2628 112
AK043248.1 - 13 - 14 2587 112
AK080701.1 - 5 - 6 741 112
AK160035.1 - 9 - 10 1571 112
BC062957.1 - 13 - 14 2645 112
DQ336396.1 - 13 - 14 2643 112

View Full Product Details