Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Pld1
Entrez Gene ID
18805
Gene Name
phospholipase D1
Gene Aliases
AA536939, C85393, Pld1a, Pld1b
Chromosome Location
Chr.3: 27938680 - 28133362 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001164056.1 NP_001157528.1 10 - 11 1170 92
NM_008875.4 NP_032901.2 9 - 10 1115 92
GenBank mRNA AK136111.1 - 4 - 5 353 92
BC068144.1 - 9 - 10 972 92
U87868.1 - 10 - 11 1153 92

View Full Product Details