Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Pld1
Entrez Gene ID
18805
Gene Name
phospholipase D1
Gene Aliases
AA536939, C85393, Pld1a, Pld1b
Chromosome Location
Chr.3: 27938680 - 28133362 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001164056.1 NP_001157528.1 7 - 8 859 104
NM_008875.4 NP_032901.2 6 - 7 804 104
GenBank mRNA BC068144.1 - 6 - 7 661 104
U87868.1 - 7 - 8 842 104

View Full Product Details