Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Mfn1
Entrez Gene ID
67414
Gene Name
mitofusin 1
Gene Aliases
2310002F04Rik, 6330416C07Rik, D3Ertd265e, HR2, mKIAA4032
Chromosome Location
Chr.3: 32529464 - 32579225 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_024200.4 NP_077162.2 16 - 17 2230 71
GenBank mRNA AK009490.1 - 1 - 2 140 71
AK018181.1 - 16 - 17 2227 71
AK081358.1 - 9 - 10 1215 71
AK141611.1 - 16 - 17 2231 71
AK208223.1 - 2 - 3 235 71
AK220169.1 - 16 - 17 2218 71
AY174062.1 - 15 - 16 2359 71
BC002133.1 - 1 - 2 199 71
BC008137.1 - 12 - 13 1497 71
BC056641.1 - 16 - 17 2189 71

View Full Product Details