Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Mfn1
Entrez Gene ID
67414
Gene Name
mitofusin 1
Gene Aliases
2310002F04Rik, 6330416C07Rik, D3Ertd265e, HR2, mKIAA4032
Chromosome Location
Chr.3: 32529464 - 32579225 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_024200.4 NP_077162.2 9 - 10 1190 92
GenBank mRNA AK081358.1 - 2 - 3 176 92
AK141611.1 - 9 - 10 1191 92
AK220169.1 - 9 - 10 1178 92
AY174062.1 - 8 - 9 1319 92
BC008137.1 - 5 - 6 457 92
BC056641.1 - 9 - 10 1149 92

View Full Product Details