Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Elf2
Entrez Gene ID
69257
Gene Name
E74-like factor 2
Gene Aliases
2610036A20Rik, A230104O07Rik, AW111824, BB183398, EU32, NERF, NERF-1A, NERF-1B, NERF-2
Chromosome Location
Chr.3: 51252720 - 51340644 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001291059.1 NP_001277988.1 2 - 3 318 137
NM_023502.2 NP_075991.1 2 - 3 318 137
GenBank mRNA AK146819.1 - 2 - 3 319 137
AK160118.1 - 2 - 3 224 137
BC027739.1 - 2 - 3 334 137
BC069901.1 - 2 - 3 314 137

View Full Product Details