The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001177732.1 | NP_001171203.1 | 16 - 17 | 2145 | 66 |
NM_001177733.1 | NP_001171204.1 | 16 - 17 | 2145 | 66 | |
NM_183191.3 | NP_899014.2 | 16 - 17 | 2145 | 66 | |
GenBank mRNA | AK173095.1 | - | 8 - 9 | 1028 | 66 |
AY691172.1 | - | 16 - 17 | 2145 | 66 | |
AY691173.1 | - | 16 - 17 | 2145 | 66 | |
AY691174.1 | - | 16 - 17 | 2145 | 66 | |
BC042549.1 | - | 4 - 5 | 474 | 66 | |
BC052372.1 | - | 4 - 5 | 474 | 66 | |
BC055005.1 | - | 16 - 17 | 2219 | 66 |