Assay Design
The assay probe spans an exon junction.
Gene Transcripts
- Gene Symbol
- Naip5
- Entrez Gene ID
-
17951
- Gene Name
- NLR family, apoptosis inhibitory protein 5
- Gene Aliases
-
Birc1e, Lgn1, Naip-rs3
- Chromosome Location
-
Chr.13: 100211739 - 100254771 on Build GRCm38
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
GenBank mRNA |
AF135493.1 |
- |
9 - 10 |
1212 |
122 |
View Full Product Details