The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_145455.3 | NP_663430.2 | 1 - 2 | 344 | 85 |
GenBank mRNA | BC064010.1 | - | 1 - 2 | 344 | 85 |