Assay Design
The assay probe spans an exon junction.
Gene Transcripts
- Gene Symbol
- Serpinb5
- Entrez Gene ID
-
20724
- Gene Name
- serine (or cysteine) peptidase inhibitor, clade B, member 5
- Gene Aliases
-
1110036M19Rik, AI462524, AI646751, Maspin, PI-5, Spi7, ovalbumin
- Chromosome Location
-
Chr.1: 106861117 - 106883350 on Build GRCm38
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
GenBank mRNA |
U54705.1 |
- |
1 - 2 |
52 |
68 |
View Full Product Details