Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Tnnt2
Entrez Gene ID
21956
Gene Name
troponin T2, cardiac
Gene Aliases
Tnt, cTnT
Chromosome Location
Chr.1: 135836316 - 135852268 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001130174.2 NP_001123646.1 11 - 12 636 78
NM_001130175.2 NP_001123647.4 11 - 12 632 78
NM_001130176.1 NP_001123648.1 9 - 10 523 78
NM_001130177.2 NP_001123649.1 10 - 11 624 78
NM_001130178.2 NP_001123650.1 10 - 11 606 78
NM_001130179.2 NP_001123651.4 10 - 11 602 78
NM_001130180.2 NP_001123652.1 9 - 10 594 78
NM_001130181.2 NP_001123653.4 9 - 10 590 78
NM_011619.3 NP_035749.1 9 - 10 594 78
GenBank mRNA AK146945.1 - 11 - 12 581 78
AK163786.1 - 10 - 11 569 78
BC063753.1 - 11 - 12 612 78
BQ554639.1 - 10 - 11 542 78
L47549.1 - 10 - 11 520 78
L47550.1 - 10 - 11 505 78
L47551.1 - 9 - 10 506 78
L47552.1 - 9 - 10 515 78
L47553.1 - 9 - 10 522 78
L47599.1 - 10 - 11 526 78
L47600.1 - 9 - 10 523 78

View Full Product Details