Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Ramp1
Entrez Gene ID
51801
Gene Name
receptor (calcitonin) activity modifying protein 1
Gene Aliases
9130218E19Rik
Chromosome Location
Chr.1: 91179822 - 91225196 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_178401.3 NP_848488.2 2 - 3 455 76
GenBank mRNA AK144331.1 - 2 - 3 455 76
BY733497.1 - 2 - 3 455 76
CB522711.1 - 1 - 2 92 76

View Full Product Details