The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001159717.1 | NP_001153189.1 | 7 - 8 | 690 | 131 |
NM_001159718.1 | NP_001153190.1 | 6 - 7 | 663 | 131 | |
NM_001159719.1 | NP_001153191.1 | 7 - 8 | 738 | 131 | |
NM_010891.2 | NP_035021.1 | 7 - 8 | 694 | 131 | |
GenBank mRNA | AK028072.1 | - | 7 - 8 | 676 | 131 |
AK146616.1 | - | 7 - 8 | 691 | 131 | |
AK151591.1 | - | 7 - 8 | 678 | 131 | |
AK169852.1 | - | 6 - 7 | 712 | 131 | |
AK171331.1 | - | 7 - 8 | 673 | 131 | |
BC138636.1 | - | 6 - 7 | 631 | 131 | |
BC138637.1 | - | 6 - 7 | 631 | 131 | |
D49382.2 | - | 7 - 8 | 649 | 131 |