Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Pou2f1
Entrez Gene ID
18986
Gene Name
POU domain, class 2, transcription factor 1
Gene Aliases
2810482H01Rik, NF-A1, Oct-1, Oct1, Otf-1, Otf1
Chromosome Location
Chr.1: 165865150 - 166002672 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_011137.3 NP_035267.2 15 - 16 1993 69
NM_198932.3 NP_945150.2 14 - 15 1921 69
NM_198933.3 NP_945151.2 14 - 15 1993 69
NM_198934.3 NP_945152.2 15 - 16 2065 69
GenBank mRNA AF508939.1 - 14 - 15 1902 69
AJ296212.1 - 14 - 15 1945 69
AK028237.1 - 15 - 16 1995 69
AK082573.1 - 2 - 3 656 69
BC066201.1 - 14 - 15 1898 69
S65461.1 - 13 - 14 1821 69
X68362.1 - 15 - 16 1966 69
X68363.1 - 14 - 15 1892 69
X68364.1 - 14 - 15 1908 69
X70324.1 - 13 - 14 1825 69
X70325.1 - 4 - 5 734 69

View Full Product Details