Assay Design
The assay probe spans an exon junction.
Gene Transcripts
- Gene Symbol
- Gin1
- Entrez Gene ID
-
252876
- Gene Name
- gypsy retrotransposon integrase 1
- Gene Aliases
-
4930429M06Rik, 8430411H10Rik, Gin-1, Zh2c2
- Chromosome Location
-
Chr.1: 97770168 - 97793710 on Build GRCm38
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
GenBank mRNA |
AK090002.1 |
- |
3 - 4 |
400 |
137 |
View Full Product Details