Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Cntnap5b
Entrez Gene ID
241175
Gene Name
contactin associated protein-like 5B
Gene Aliases
C230078M14Rik, Caspr5-2
Chromosome Location
Chr.1: 99772247 - 100486066 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_172851.2 NP_766439.2 20 - 21 3309 84
GenBank mRNA AK082656.1 - 13 - 14 2196 84
AK133394.1 - 10 - 11 1639 84
BC145618.1 - 13 - 14 2074 84

View Full Product Details