Assay Design
The assay probe spans an exon junction.
Gene Transcripts
- Gene Symbol
- Dcaf8
- Entrez Gene ID
-
98193
- Gene Name
- DDB1 and CUL4 associated factor 8
- Gene Aliases
-
AA408877, AA433120, AA792214, AU018973, C76055, D1Dau35e, D1Ucla4, H326, Wdr42a
- Chromosome Location
-
Chr.1: 172148015 - 172196393 on Build GRCm38
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
GenBank mRNA |
BC023704.1 |
- |
11 - 12 |
1553 |
107 |
View Full Product Details