Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Sctr
Entrez Gene ID
319229
Gene Name
secretin receptor
Gene Aliases
6530402O03Rik
Chromosome Location
Chr.1: 120006891 - 120063549 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001012322.2 NP_001012322.2 1 - 2 354 62
GenBank mRNA AK032652.1 - 1 - 2 356 62
BC089355.1 - 1 - 2 300 62

View Full Product Details