Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Atp1a4
Entrez Gene ID
27222
Gene Name
ATPase, Na+/K+ transporting, alpha 4 polypeptide
Gene Aliases
-
Chromosome Location
Chr.1: 172223508 - 172258424 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_013734.1 NP_038762.1 18 - 19 2698 82

View Full Product Details