Assay Design
The assay probe spans an exon junction.
Gene Transcripts
- Gene Symbol
- Ilkap
- Entrez Gene ID
-
67444
- Gene Name
- integrin-linked kinase-associated serine/threonine phosphatase 2C
- Gene Aliases
-
0710007A14Rik, 1600009O09Rik, PP2C-DELTA
- Chromosome Location
-
Chr.1: 91373846 - 91398841 on Build GRCm38
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
GenBank mRNA |
AK005408.1 |
- |
4 - 5 |
406 |
63 |
BC027439.1 |
- |
4 - 5 |
272 |
63 |
View Full Product Details