Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Agap1
Entrez Gene ID
347722
Gene Name
ArfGAP with GTPase domain, ankyrin repeat and PH domain 1
Gene Aliases
Centg2, Ggap1, R75364, mKIAA1099
Chromosome Location
Chr.1: 89454811 - 89895282 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001037136.1 NP_001032213.1 7 - 8 1422 76
NM_178119.3 NP_835220.1 7 - 8 1422 76
GenBank mRNA AK046523.1 - 7 - 8 1383 76
AK129289.2 - 7 - 8 1369 76
AK147503.1 - 7 - 8 1428 76
AK147521.1 - 8 - 9 2073 76
BC096588.1 - 7 - 8 1370 76

View Full Product Details