The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_021353.3 | NP_067328.1 | 14 - 15 | 1773 | 100 |
GenBank mRNA | AF136751.1 | - | 14 - 15 | 1691 | 100 |
BC139271.1 | - | 13 - 14 | 1608 | 100 | |
BC139273.1 | - | 13 - 14 | 1608 | 100 |