Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Sgcd
Entrez Gene ID
24052
Gene Name
sarcoglycan, delta (dystrophin-associated glycoprotein)
Gene Aliases
35kDa, delta-SG
Chromosome Location
Chr.11: 46971077 - 47988385 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
GenBank mRNA AK047384.1 - - 899 92
AK048480.1 - 2 - 3 240 92

View Full Product Details