Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Lhx9
Entrez Gene ID
16876
Gene Name
LIM homeobox protein 9
Gene Aliases
3110009O07Rik, BB104635, LH2B
Chromosome Location
Chr.1: 138825186 - 138848649 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_010714.3 NP_034844.1 2 - 3 145 108
GenBank mRNA AK013209.1 - 1 - 2 171 108

View Full Product Details