The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001163570.1 | NP_001157042.1 | 1 - 2 | 355 | 87 |
NM_027148.3 | NP_081424.3 | 1 - 2 | 355 | 87 | |
GenBank mRNA | AK047257.1 | - | 1 - 2 | 358 | 87 |
AK220227.1 | - | 1 - 2 | 355 | 87 | |
BC059089.1 | - | 1 - 2 | 384 | 87 |