Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Mbnl1
Entrez Gene ID
56758
Gene Name
muscleblind-like 1 (Drosophila)
Gene Aliases
Mbnl, mKIAA0428
Chromosome Location
Chr.3: 60472791 - 60629750 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001253711.1 NP_001240640.1 6 - 7 1008 66
GenBank mRNA BC060031.1 - 7 - 8 2035 66

View Full Product Details