The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_175191.4 | NP_780400.2 | 2 - 3 | 1418 | 96 |
GenBank mRNA | AK034194.1 | - | 2 - 3 | 1372 | 96 |
AK048077.1 | - | 2 - 3 | 1373 | 96 | |
AK140577.1 | - | 2 - 3 | 1359 | 96 | |
AK141569.1 | - | 2 - 3 | 1403 | 96 | |
BC089509.1 | - | 2 - 3 | 1419 | 96 |